Novus Biologicals
Manufacturer Code:NBP159514
Catalog # NBP159514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC26A9(solute carrier family 26 member 9) The peptide sequence was selected from the middle region of SLC26A9. Peptide sequence LAKLSSTYGKIGVKVFLVNIHAQVYNDISHGGVFEDGSLECKHVFPSIHD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anion transporter/exchanger protein 9; Anion transporter/exchanger protein 9 anion transporter/exchanger-9 solute carrier family 26 member 9 solute carrier family 26 member 9; anion transporter/exchanger-9; solute carrier family 26 (anion exchanger), member 9; Solute carrier family 26 member 9; solute carrier family 26, member 9
Gene Aliases: SLC26A9
UniProt ID: (Human) Q7LBE3
Entrez Gene ID: (Human) 115019
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.