Invitrogen
Manufacturer Code:MA146406
Catalog # PIMA146406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 14H5 |
Immunogen | Fusion protein containing the C-terminal region of mouse SLC26A7. &lsqbSwiss-ProtNo. Q24JS8] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anion exchange transporter; anion exchanger Slc26a7; RP23-272I16.1 MGC30955; slc26a7; Solute carrier family 26 member 7
Gene Aliases: Slc26a7
UniProt ID: (Mouse) Q8R2Z3
Entrez Gene ID: (Mouse) 208890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.