Novus Biologicals
Manufacturer Code:NB11060965AF405
Catalog # NB11060965W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 14H5 |
Immunogen | A fusion protein containing the C-terminal region of mouse SLC26A7. [Swiss-Prot# Q24JS8] |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anion exchange transporter; MGC126268 solute carrier family 26 member 7 sulfate anion transporter; SLC26A7; solute carrier family 26 (anion exchanger), member 7; Solute carrier family 26 member 7; solute carrier family 26, member 7; sulfate anion transporter
Gene Aliases: SLC26A7; SUT2
UniProt ID: (Human) Q8TE54
Entrez Gene ID: (Human) 115111
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.