Invitrogen
Manufacturer Code:PA561722
Catalog # PIPA561722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC26A6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anion exchange transporter; anion transporter 1; pendrin L1; Pendrin-L1; Pendrin-like protein 1; solute carrier family 26 (anion exchanger), member 6; Solute carrier family 26 member 6; solute carrier family 26 member 6 DKFZp586E1422 anion transporter 1 pendrin L1 pendrin-like protein 1 sulfate anion transporter; sulfate anion transporter
Gene Aliases: SLC26A6
UniProt ID: (Human) Q9BXS9
Entrez Gene ID: (Human) 65010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.