Invitrogen
Manufacturer Code:PA537971
Catalog # PIPA537971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RDVAEYSEAKEVR from the internal region of SLC26A6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Anion exchange transporter; anion transporter 1; Chloride-formate exchanger; Pat-1; pendrin L1; Pendrin-L1; Pendrin-like protein 1; Putative anion transporter-1; solute carrier family 26 (anion exchanger), member 6; Solute carrier family 26 member 6; solute carrier family 26 member 6 DKFZp586E1422 anion transporter 1 pendrin L1 pendrin-like protein 1 sulfate anion transporter; sulfate anion transporter
Gene Aliases: B930010B04Rik; CFEX; Pat-1; Pat1; SLC26A6
UniProt ID: (Human) Q9BXS9, (Mouse) Q8CIW6
Entrez Gene ID: (Human) 65010, (Mouse) 171429
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.