Invitrogen
Manufacturer Code:PA5114178
Catalog # PIPA5114178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: VNTSLEDMRS NNVEDCKMMV SSGDKMEDAT ANGQEDSKAP DGSTLKALGL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anion exchange transporter; Anion exchange transporter anion transporter 1 B930010B04Rik Cfex Chloride-formate exchanger DKFZp586E1422 Pat1 Pat-1 pendrin L1 Pendrin-L1 Pendrin-like protein 1 Putative anion transporter-1 Slc26a6 solute carrier family 26 (anion exchanger) member 6 Solute carrier family 26 member 6 solute carrier family 26 member 6 sulfate anion transporter; anion transporter 1; pendrin L1; Pendrin-L1; Pendrin-like protein 1; solute carrier family 26 (anion exchanger), member 6; Solute carrier family 26 member 6; sulfate anion transporter
Gene Aliases: SLC26A6
UniProt ID: (Human) Q9BXS9
Entrez Gene ID: (Human) 65010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.