Abnova
Manufacturer Code:H00065010B01P
Catalog # 89-017-161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC26A6 (NP_599025.2 1 a.a. ~ 758 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SLC26A6 protein.
For Research Use Only
Protein Aliases: Anion exchange transporter; anion transporter 1; DKFZp586E1422; pendrin L1; Pendrin-L1; Pendrin-like protein 1; solute carrier family 26 (anion exchanger), member 6; Solute carrier family 26 member 6; sulfate anion transporter
Gene Aliases: SLC26A6
UniProt ID: (Human) Q9BXS9
Entrez Gene ID: (Human) 65010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.