Invitrogen
Manufacturer Code:PA542533
Catalog # PIPA542533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC26A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: outer hair cell motor protein; Prestin; prestin (motor protein); solute carrier family 26 (anion exchanger), member 5; Solute carrier family 26 member 5; Solute carrier family 26 member 5 (prestin) DFNB61 MGC118886 MGC118887 MGC118888 MGC118889 PRES; solute carrier family 26, member 5 (prestin); solute carrier family 26, member 5 (prestin) isoform 1; solute carrier family 26, member 5 (prestin) isoform 2
Gene Aliases: DFNB61; PRES; prestin; SLC26A5
UniProt ID: (Human) P58743, (Mouse) Q99NH7, (Rat) Q9EPH0
Entrez Gene ID: (Rabbit) 100216333, (Horse) 100058502, (Dog) 483274, (Human) 375611, (Bovine) 536341, (Mouse) 80979, (Guinea pig) 100727496, (Rat) 83819
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.