Invitrogen
Manufacturer Code:PA5115911
Catalog # PIPA5115911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC26A4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DFNB4 EVA PDS Pendred syndrome homolog Pendred's syndrome pendrin SLC26A4 sodium-independent chloride/iodide transporter solute carrier family 26 (anion exchanger) member 4 solute carrier family 26 member 4 solute carrier family 26 member 4 TDH2B truncated solute carrier family 26; Pendred syndrome homolog; Pendred's syndrome; Pendrin; Sodium-independent chloride/iodide transporter; solute carrier family 26 (anion exchanger), member 4; Solute carrier family 26 member 4; solute carrier family 26, member 4; truncated solute carrier family 26
Gene Aliases: DFNB4; EVA; PDS; pendrin; SLC26A4; TDH2B
UniProt ID: (Human) O43511, (Rat) Q9R154, (Mouse) Q9R155
Entrez Gene ID: (Human) 5172, (Rat) 29440, (Mouse) 23985
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.