Invitrogen
Manufacturer Code:PA568530
Catalog # PIPA568530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SLC26A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anion exchange protein; Chloride anion exchanger; CLD DRA; Down-regulated in adenoma; down-regulated in adenoma DRA; down-regulated in adenoma protein; Protein DRA; solute carrier family 26 (anion exchanger), member 3; Solute carrier family 26 member 3; solute carrier family 26, member 3
Gene Aliases: 9030623B18Rik; 9130013M11Rik; AV376035; CLD; DRA; SLC26A3
UniProt ID: (Human) P40879, (Mouse) Q9WVC8, (Rat) Q924C9
Entrez Gene ID: (Human) 1811, (Sheep) 100462650, (Rabbit) 100008798, (Bovine) 512856, (Pig) 100170644, (Dog) 483261, (Mouse) 13487, (Rat) 114629, (Guinea pig) 100689262
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.