Novus Biologicals
Manufacturer Code:NBP30507920UL
Catalog # NB096279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 505-764 of human SLC26A3 (NP_000102.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chloride anion exchanger; chloride anion exchanger CLD congenital chloride diarrhea down-regulated in adenoma protein DRADown-regulated in adenoma Protein DRA Solute carrier family 26 member 3 solute carrier family 26 member 3; Down-regulated in adenoma; down-regulated in adenoma protein; Protein DRA; solute carrier family 26 (anion exchanger), member 3; Solute carrier family 26 member 3; solute carrier family 26, member 3
Gene Aliases: CLD; DRA; SLC26A3
UniProt ID: (Human) P40879
Entrez Gene ID: (Human) 1811
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.