Abnova
Manufacturer Code:H00001811A01
Catalog # 89-104-396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 503 a.a. &bsim 600 a.a) partial recombinant protein with GST tag.,SLC26A3 (NP_000102 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLC26A3.
For Research Use Only
Protein Aliases: Chloride anion exchanger; CLD; Down-regulated in adenoma; down-regulated in adenoma protein; DRA; Protein DRA; solute carrier family 26 (anion exchanger), member 3; Solute carrier family 26 member 3; solute carrier family 26, member 3
Gene Aliases: CLD; DRA; SLC26A3
UniProt ID: (Human) P40879
Entrez Gene ID: (Human) 1811
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.