Invitrogen
Manufacturer Code:PA569251
Catalog # PIPA569251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse SLC26A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diastrophic dysplasia protein; Diastrophic dysplasia protein homolog; DTD sulfate transporter; solute carrier family 26 (anion exchanger), member 2; solute carrier family 26 (sulfate transporter), member 2; Solute carrier family 26 member 2; solute carrier protein 26a2; ST-OB; sulfate anion transporter 1; Sulfate transporter
Gene Aliases: D5S1708; DTD; DTDST; EDM4; MST153; MSTP157; SLC26A2; ST-OB
UniProt ID: (Bovine) Q9BEG8, (Sheep) Q9GJY3, (Horse) Q65AC2, (Human) P50443, (Mouse) Q62273, (Rat) O70531
Entrez Gene ID: (Bovine) 513953, (Sheep) 100294558, (Horse) 100034141, (Dog) 612273, (Human) 1836, (Rabbit) 100342617, (Guinea pig) 100725509, (Mouse) 13521, (Rat) 117267
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.