Invitrogen
Manufacturer Code:PA5116742
Catalog # PIPA5116742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 500-600 of human SLC26A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anion exchanger Slc26a11; novel sulfate transporter protein (Slc26a11); SLC26A11; SLC26A11 Sodium-independent sulfate anion transporter solute carrier family 26 (anion exchanger) member 11 Solute carrier family 26 member 11 solute carrier family 26 member 11; Sodium-independent sulfate anion transporter; solute carrier family 26 (anion exchanger), member 11; Solute carrier family 26 member 11; solute carrier family 26, member 11
Gene Aliases: F630021I08Rik; SLC26A11
UniProt ID: (Human) Q86WA9, (Mouse) Q80ZD3
Entrez Gene ID: (Human) 284129, (Mouse) 268512
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.