Novus Biologicals
Manufacturer Code:NBP297794AF488
Catalog # NB047845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SLC26A11 fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC46523 sodium-independent sulfate anion transporter Solute carrier family 26 member 11 solute carrier family 26 member 11; SLC26A11; Sodium-independent sulfate anion transporter; solute carrier family 26 (anion exchanger), member 11; Solute carrier family 26 member 11; solute carrier family 26, member 11
Gene Aliases: SLC26A11
UniProt ID: (Human) Q86WA9
Entrez Gene ID: (Human) 284129
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.