Novus Biologicals
Manufacturer Code:NBP220393
Catalog # NBP220393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human SLC25A6. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenine nucleotide translocator 3; Adenine nucleotide translocator 3 ADP ADP/ATP translocase 3 ADP/ATP translocator of liver ANT 2 ANT 3 ANT3AAC3 ANT3Y ATP carrier protein ATP carrier protein 3 ATP carrier protein isoform T2 ATP carrier protein liver MGC17525 solute carrier family 25 (mitochondrial carrier adenine nucleotidetranslocator) member 6 Solute carrier family 25 member 6; ADP,ATP carrier protein 3; ADP,ATP carrier protein, isoform T2; ADP,ATP carrier protein, liver; ADP/ATP translocase 3; ADP/ATP translocase 3, N-terminally processed; ADP/ATP translocator of liver; ANT 2; ANT 3; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6; Solute carrier family 25 member 6
Gene Aliases: AAC3; ANT; ANT 2; ANT 3; ANT3; ANT3Y; CDABP0051; SLC25A6
UniProt ID: (Human) P12236
Entrez Gene ID: (Human) 293
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.