Invitrogen
Manufacturer Code:PA568086
Catalog # PIPA568086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human SLC25A5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenine nucleotid translocator 2 fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotid translocator 2, fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); adenine nucleotide translocator 2 fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocator 2, fibroblast isoform (ATP-ADP carrier protein); ADP,ATP carrier protein 2; ADP,ATP carrier protein, fibroblast isoform; ADP/ATP translocase 2; ADP/ATP translocase 2 ADPATP carrier protein 2 ADPATP carrier protein fibroblast isoform Adenine nucleotide translocator 2 ANT 2 Solute carrier family 25 member 5 DP/ATP translocase 2 N-terminally processed ANT2; ADP/ATP translocase 2, N-terminally processed; ANT 2; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; Solute carrier family 25 member 5
Gene Aliases: 2F1; AAC2; ANT2; SLC25A5; T2; T3
UniProt ID: (Human) P05141, (Rat) Q09073, (Mouse) P51881
Entrez Gene ID: (Human) 292, (Rat) 25176, (Mouse) 11740
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.