Abnova
Manufacturer Code:H00000292K
Catalog # 89-208-337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC25A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC25A5 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 2F1 AAC2 ANT2 T2 T3; Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); ADP,ATP carrier protein 2; ADP,ATP carrier protein, fibroblast isoform; ADP/ATP translocase 2; ADP/ATP translocase 2, N-terminally processed; ANT 2; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; Solute carrier family 25 member 5
Gene Aliases: 2F1; AAC2; ANT2; SLC25A5; T2; T3
UniProt ID: (Human) P05141
Entrez Gene ID: (Human) 292
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.