Proteintech
Manufacturer Code:278691AP150UL
Catalog # 50-173-5779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC25A46 Fusion Protein Ag27293 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SLC25A46 TB1; Solute carrier family 25 member 46; solute carrier family 25, member 46
Gene Aliases: 1200007B05Rik; AI325987; HMSN6B; SLC25A46; TB1
UniProt ID: (Human) Q96AG3, (Mouse) Q9CQS4
Entrez Gene ID: (Human) 91137, (Mouse) 67453
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.