Invitrogen
Manufacturer Code:PA542467
Catalog # PIPA542467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SLC25A39 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Solute carrier family 25 member 39
Gene Aliases: 3010027G13Rik; CGI-69; CGI69; D11Ertd333e; PRO2163; RGD1306193; SLC25A39; zgc:63736
UniProt ID: (Human) Q9BZJ4, (Bovine) Q17QI7, (Rat) Q4V8K4, (Mouse) Q9D8K8, (Zebrafish) Q7SXW0
Entrez Gene ID: (Horse) 100064639, (Dog) 490938, (Rabbit) 100338521, (Human) 51629, (Bovine) 509684, (Guinea pig) 100728216, (Rat) 360636, (Mouse) 68066, (Zebrafish) 393458
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.