Invitrogen
Manufacturer Code:PA561068
Catalog # PIPA561068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC25A37 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MFRN MFRN1 MSC MSCP PRO1278 PRO1584 PRO2217 mitochondrial iron transporter 1 mitochondrial solute carrier protein mitoferrin mitoferrin-1 predicted protein of HQ2217; Mitochondrial iron transporter 1; Mitochondrial solute carrier protein; mitoferrin; Mitoferrin-1; predicted protein of HQ2217; solute carrier family 25 (mitochondrial iron transporter), member 37; Solute carrier family 25 member 37; solute carrier family 25, member 37
Gene Aliases: HT015; MFRN; MFRN1; MSC; MSCP; PRO1278; PRO1584; PRO2217; SLC25A37
UniProt ID: (Human) Q9NYZ2
Entrez Gene ID: (Human) 51312
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.