Abnova
Manufacturer Code:PAB24440
Catalog # 89-126-156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC25A37. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC25A37.
For Research Use Only
Protein Aliases: HT015; MFRN; Mitochondrial iron transporter 1; Mitochondrial solute carrier protein; mitoferrin; Mitoferrin-1; MSC; MSCP; predicted protein of HQ2217; PRO1278; PRO1584; PRO2217; solute carrier family 25 (mitochondrial iron transporter), member 37; Solute carrier family 25 member 37; solute carrier family 25, member 37
Gene Aliases: HT015; MFRN; MFRN1; MSC; MSCP; PRO1278; PRO1584; PRO2217; SLC25A37
UniProt ID: (Human) Q9NYZ2
Entrez Gene ID: (Human) 51312
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.