Novus Biologicals
Manufacturer Code:NBP15949720UL
Catalog # NBP15949720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC25A36(solute carrier family 25 member 36) The peptide sequence was selected from the N terminal of SLC25A36. Peptide sequence SSSVTLYISEVQLNTMAGASVNRVVSPGPLHCLKVILEKEGPRSLFRGLG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp564C053 FLJ10618 solute carrier family 25 member 36 solute carrier family 25 member 36; solute carrier family 25 (pyrimidine nucleotide carrier ), member 36; solute carrier family 25 (pyrimidine nucleotide carrier), member 36; Solute carrier family 25 member 36; solute carrier family 25, member 36
Gene Aliases: PNC2; SLC25A36
UniProt ID: (Human) Q96CQ1
Entrez Gene ID: (Human) 55186
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.