Novus Biologicals
Manufacturer Code:NBP2857400.1ML
Catalog # NB033669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human SLC25A33 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMSC-MCP; BMSC-MCPPNC1 protein Bone marrow stromal cell mitochondrial carrier protein HuBMSC-MCP MGC4399 mitochondrial carrier protein novel mitochondrial carrier protein Protein PNC1 solute carrier family 25 member 33 solute carrier family 25 member 33; Bone marrow stromal cell mitochondrial carrier protein; huBMSC-MCP; mitochondrial carrier protein; novel mitochondrial carrier protein; PNC1 protein; Protein PNC1; solute carrier family 25 (pyrimidine nucleotide carrier), member 33; Solute carrier family 25 member 33; solute carrier family 25, member 33
Gene Aliases: BMSC-MCP; PNC1; SLC25A33
UniProt ID: (Human) Q9BSK2
Entrez Gene ID: (Human) 84275
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.