Invitrogen
Manufacturer Code:PA544133
Catalog # PIPA544133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC25A31 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenine nucleotide translocase 4; Adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 4; ADP/ATP translocase 4; ANT 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; Solute carrier family 25 member 31; Sperm flagellar energy carrier protein
Gene Aliases: 1700034J06Rik; AAC4; ANT 4; ANT4; SFEC; SFEC35kDa; SLC25A31
UniProt ID: (Human) Q9H0C2, (Bovine) Q2YDD9, (Mouse) Q3V132
Entrez Gene ID: (Human) 83447, (Bovine) 541168, (Rabbit) 100346477, (Dog) 483832, (Horse) 100071908, (Guinea pig) 100722295, (Rat) 689108, (Mouse) 73333
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.