Novus Biologicals
Manufacturer Code:NBP286812
Catalog # NB034434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human SLC25A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PHCmitochondrial phosphate carrier protein phosphate carrier protein mitochondrial Phosphate transport protein PTP solute carrier family 25 (mitochondrial carrier phosphate carrier) member 3 Solute carrier family 25 member 3; Phosphate carrier protein, mitochondrial; Phosphate transport protein; PTP; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 3; Solute carrier family 25 member 3
Gene Aliases: OK/SW-cl.48; PHC; PTP; SLC25A3
UniProt ID: (Human) Q00325
Entrez Gene ID: (Human) 5250
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.