Proteintech
Manufacturer Code:158551AP150UL
Catalog # 50-173-0532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC25A3 Fusion Protein Ag8698 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OK/SW cl.48 PHC phosphate carrier Phosphate transport protein PiC PTP SLC25A3; Phosphate carrier protein, mitochondrial; phosphate carrier, mitochondrial; Phosphate transport protein; PTP; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 3; Solute carrier family 25 member 3
Gene Aliases: 5730556H19Rik; OK/SW-cl.48; PHC; PTP; SLC25A3
UniProt ID: (Human) Q00325, (Mouse) Q8VEM8
Entrez Gene ID: (Human) 5250, (Mouse) 18674
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.