Invitrogen
Manufacturer Code:PA542417
Catalog # PIPA542417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SLC25A29 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CACT-like; carnitine-acylcarnitine translocase like; Carnitine/acylcarnitine translocase-like; mitchondrial carnitine/acylcarnitine carrier protein CACL; Mitochondrial basic amino acids transporter; Mitochondrial carnitine/acylcarnitine carrier protein CACL; Mitochondrial ornithine transporter 3; solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29; solute carrier family 25 (mitochondrial carrier, palmitoylcarnitine transporter), member 29; Solute carrier family 25 member 29; solute carrier family 25, member 29
Gene Aliases: C030003J19Rik; C14orf69; CACL; mCACL; ORNT3; SLC25A29; zgc:114094
UniProt ID: (Human) Q8N8R3, (Bovine) Q08DK7, (Mouse) Q8BL03, (Rat) Q5HZE0
Entrez Gene ID: (Guinea pig) 100715169, (Rabbit) 100349627, (Horse) 100050099, (Human) 123096, (Bovine) 507686, (Dog) 490856, (Mouse) 214663, (Rat) 314441, (Zebrafish) 569608
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.