Invitrogen
Manufacturer Code:PA569287
Catalog # PIPA569287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human SLC25A26 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; SAMC; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; Solute carrier family 25 member 26; solute carrier family 25, member 26; SZFULLNAME2
Gene Aliases: 4930433D19Rik; 4933433F13Rik; AW557176; COXPD28; D6Bwg0781e; SAMC; SLC25A26; Slc25a6; SZGENESYMBOL2; zgc:110080
UniProt ID: (Bovine) A6QR09, (Human) Q70HW3, (Mouse) Q5U680, (Zebrafish) Q4V9P0
Entrez Gene ID: (Dog) 611050, (Sheep) 100147801, (Bovine) 614713, (Pig) 100624317, (Human) 115286, (Rabbit) 100338902, (Mouse) 67582, (Rat) 362403, (Zebrafish) 560478
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.