Invitrogen
Manufacturer Code:PA555452
Catalog # PIPA555452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC25A26 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial S-adenosylmethionine transporter; S-adenosylmethionine mitochondrial carrier protein; SAMC; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; Solute carrier family 25 member 26
Gene Aliases: COXPD28; SAMC; SLC25A26
UniProt ID: (Human) Q70HW3
Entrez Gene ID: (Human) 115286
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.