Invitrogen
Manufacturer Code:PA542459
Catalog # PIPA542459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC25A24 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-binding mitochondrial carrier protein SCaMC-1; calcium-binding mitochondrial carrier protein SCaMC-1-like; Calcium-binding mitochondrial carrier protein SCaMC-1-like protein; calcium-binding transporter; Mitochondrial ATP-Mg/Pi carrier protein 1; Mitochondrial Ca(2+)-dependent solute carrier protein 1; Peroxisomal Ca(2+)-dependent solute carrier; peroxisomal Ca-dependent solute carrier; short calcium-binding mitochondrial carrier 1; similar to mitochondrial solute carrier; Small calcium-binding mitochondrial carrier protein 1; Solute carrier family 25 (mitochondrial carrier phosphate carrier) member 24 RP11-356N1.3 APC1 DKFZp586G0123 SCAMC-1; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24; Solute carrier family 25 member 24
Gene Aliases: 2610016M12Rik; APC1; EFINAL; MCSC1; SCAMC-1; SCAMC1; si:ch211-63o20.9; SLC25A24; zgc:92470
UniProt ID: (Bovine) A5PJZ1, (Rabbit) O18757, (Human) Q6NUK1, (Mouse) Q8BMD8, (Zebrafish) Q66L49
Entrez Gene ID: (Horse) 100057835, (Bovine) 534742, (Rabbit) 100009567, (Human) 29957, (Mouse) 229731, (Rat) 310791, (Zebrafish) 447867
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.