Abnova
Manufacturer Code:H00029957K
Catalog # 89-202-270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC25A24 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC25A24 peptide using ARM Technology.
For Research Use Only
Protein Aliases: APC1 DKFZp586G0123 SCAMC-1; Calcium-binding mitochondrial carrier protein SCaMC-1; calcium-binding transporter; Mitochondrial ATP-Mg/Pi carrier protein 1; Mitochondrial Ca(2+)-dependent solute carrier protein 1; short calcium-binding mitochondrial carrier 1; Small calcium-binding mitochondrial carrier protein 1; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 24; Solute carrier family 25 member 24
Gene Aliases: APC1; MCSC1; SCAMC-1; SCAMC1; SLC25A24
UniProt ID: (Human) Q6NUK1
Entrez Gene ID: (Human) 29957
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.