Invitrogen
Manufacturer Code:PA564930
Catalog # PIPA564930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human SLC25A23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APC2 MCSC2 SCaMC-3 calcium-binding mitochondrial carrier protein SCaMC-3 mitochondrial ATP-Mg/Pi carrier protein 2 mitochondrial Ca(2+)-dependent solute carrier protein 2 mitochondrial Ca2+-dependent solute carrier protein 2 short calcium-binding mitochondrial carrier 3 sma; Calcium-binding mitochondrial carrier protein SCaMC-3; Mitochondrial ATP-Mg/Pi carrier protein 2; Mitochondrial Ca(2+)-dependent solute carrier protein 2; mitochondrial Ca2+-dependent solute carrier protein 2; short calcium-binding mitochondrial carrier 3; Small calcium-binding mitochondrial carrier protein 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23; Solute carrier family 25 member 23
Gene Aliases: APC2; MCSC2; SCaMC-3; SCAMC3; SLC25A23
UniProt ID: (Human) Q9BV35
Entrez Gene ID: (Human) 79085
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.