Invitrogen
Manufacturer Code:PA5116578
Catalog # PIPA5116578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human SLC25A23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310067G05Rik APC2 Calcium-binding mitochondrial carrier protein SCaMC-3 MCSC2 Mitochondrial ATP-Mg/Pi carrier protein 2 mitochondrial Ca(2+)-dependent solute carrier protein 2 mitochondrial Ca2+-dependent solute carrier protein 2 phosphate carrier) member 23 SCAMC3 SCaMC-3 short calcium-binding mitochondrial carrier 3 Slc25a23 sma small calcium-binding mitochondrial carrier protein 3 solute carrier family 25 (mitochondrial carrier phosphate carrier) member 23 solute carrier family 25 member 23; Calcium-binding mitochondrial carrier protein SCaMC-3; Mitochondrial ATP-Mg/Pi carrier protein 2; Mitochondrial Ca(2+)-dependent solute carrier protein 2; mitochondrial Ca2+-dependent solute carrier protein 2; phosphate carrier), member 23; short calcium-binding mitochondrial carrier 3; Small calcium-binding mitochondrial carrier protein 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23; Solute carrier family 25 member 23
Gene Aliases: 2310067G05Rik; APC2; MCSC2; SCaMC-3; SCAMC3; SLC25A23
UniProt ID: (Human) Q9BV35, (Mouse) Q6GQS1
Entrez Gene ID: (Human) 79085, (Rat) 301113, (Mouse) 66972
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.