Invitrogen
Manufacturer Code:PA542492
Catalog # PIPA542492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC25A22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GC-1; Glutamate/H(+) symporter 1; Mitochondrial glutamate carrier 1; solute carrier family 25 (mitochondrial carrier, glutamate), member 22; Solute carrier family 25 (mitochondrial carrier: glutamate) member 22 FLJ13044 GC1 EIEE3 NET44; solute carrier family 25 (mitochondrial carrier: glutamate), member 22; Solute carrier family 25 member 22
Gene Aliases: 1300006L01Rik; AI060884; EIEE3; GC-1; GC1; NET44; RGD1307826; SLC25A22
UniProt ID: (Human) Q9H936, (Bovine) Q08DK4, (Mouse) Q9D6M3
Entrez Gene ID: (Horse) 100147341, (Human) 79751, (Pig) 100514030, (Bovine) 504371, (Mouse) 68267, (Guinea pig) 100717225, (Rat) 309111
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.