Novus Biologicals
Manufacturer Code:NBP159603
Catalog # NBP159603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC25A22(solute carrier family 25 (mitochondrial carrier: glutamate) member 22) The peptide sequence was selected from the N terminal of SLC25A22. Peptide sequence LQDAGRIAAQRKILAAQGQLSAQGGAQPSVEAPAAPRP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EIEE3mitochondrial glutamate carrier 1 FLJ13044 GC1GC-1 Glutamate/H(+) symporter 1 NET44 solute carrier family 25 (mitochondrial carrier: glutamate) member 22 Solute carrier family 25 member 22; GC-1; Glutamate/H(+) symporter 1; Mitochondrial glutamate carrier 1; solute carrier family 25 (mitochondrial carrier: glutamate), member 22; Solute carrier family 25 member 22
Gene Aliases: EIEE3; GC-1; GC1; NET44; SLC25A22
UniProt ID: (Human) Q9H936
Entrez Gene ID: (Human) 79751
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.