Invitrogen
Manufacturer Code:PA569279
Catalog # PIPA569279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SLC25A21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial 2-oxodicarboxylate carrier; mitochondrial solute carrier family 25 member 21; oxodicarboxylate carrier; Solute carrier family 25 (mitochondrial oxoadipate carrier) member 21 MGC126570 ODC ODC1; solute carrier family 25 (mitochondrial oxoadipate carrier), member 21; solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21; Solute carrier family 25 member 21
Gene Aliases: 9930033G19Rik; A630030I10; BB158148; ODC; ODC1; SLC25A21; zgc:153387
UniProt ID: (Human) Q9BQT8, (Bovine) A0JN87, (Rat) Q99JD3, (Mouse) Q8BZ09
Entrez Gene ID: (Human) 89874, (Dog) 490655, (Bovine) 513423, (Rabbit) 103351195, (Horse) 100059414, (Guinea pig) 100730196, (Rat) 171151, (Mouse) 217593, (Zebrafish) 767694
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.