Abnova
Manufacturer Code:PAB17768
Catalog # 89-119-435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human SLC25A21. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC25A21.
For Research Use Only
Protein Aliases: MGC126570; Mitochondrial 2-oxodicarboxylate carrier; ODC; ODC1; oxodicarboxylate carrier; solute carrier family 25 (mitochondrial oxoadipate carrier), member 21; solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21; Solute carrier family 25 member 21
Gene Aliases: ODC; ODC1; SLC25A21
UniProt ID: (Human) Q9BQT8
Entrez Gene ID: (Human) 89874
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.