Invitrogen
Manufacturer Code:PA542411
Catalog # PIPA542411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SLC25A20 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAC; carnitine/acylcarnitine carrier protein; Carnitine/acylcarnitine translocase; mitochondrial carnitine-acylcarnitine translocase; Mitochondrial carnitine/acylcarnitine carrier protein; Solute carrier family 25 (carnitine/acylcarnitine translocase) member 20 CAC CACT; solute carrier family 25 (carnitine/acylcarnitine translocase), member 20; solute carrier family 25 (mitochondrial carnitine/acylcarnitine translocase), member 20; Solute carrier family 25 member 20
Gene Aliases: 1110007P09Rik; C78826; CAC; CACT; mCAC; SLC25A20; zgc:77760
UniProt ID: (Human) O43772, (Mouse) Q9Z2Z6
Entrez Gene ID: (Human) 788, (Bovine) 518758, (Rabbit) 100352079, (Sheep) 100147786, (Dog) 484774, (Horse) 100063527, (Guinea pig) 100729199, (Rat) 117035, (Mouse) 57279, (Zebrafish) 393833
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.