Proteintech
Manufacturer Code:193631AP150UL
Catalog # 50-173-2389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC25A20 Fusion Protein Ag6896 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAC; CAC CACT SLC25A20; Carnitine/acylcarnitine translocase; mitochondrial carnitine-acylcarnitine translocase; Mitochondrial carnitine/acylcarnitine carrier protein; solute carrier family 25 (carnitine/acylcarnitine translocase), member 20; solute carrier family 25 (mitochondrial carnitine/acylcarnitine translocase), member 20; Solute carrier family 25 member 20
Gene Aliases: 1110007P09Rik; C78826; CAC; CACT; mCAC; SLC25A20
UniProt ID: (Human) O43772, (Mouse) Q9Z2Z6
Entrez Gene ID: (Human) 788, (Rat) 117035, (Mouse) 57279
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.