Abnova
Manufacturer Code:H00000788K
Catalog # 89-208-318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC25A20 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC25A20 peptide using ARM Technology.
For Research Use Only
Protein Aliases: CAC; CAC CACT; Carnitine/acylcarnitine translocase; Mitochondrial carnitine/acylcarnitine carrier protein; solute carrier family 25 (carnitine/acylcarnitine translocase), member 20; Solute carrier family 25 member 20
Gene Aliases: CAC; CACT; SLC25A20
UniProt ID: (Human) O43772
Entrez Gene ID: (Human) 788
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.