Invitrogen
Manufacturer Code:PA570702
Catalog # PIPA570702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human SLC25A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mitochondrial ornithine transporter; Mitochondrial ornithine transporter 2; ornithine transporter 2; ORNT2; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 2; Solute carrier family 25 member 2
Gene Aliases: ORC2; ORNT2; SLC25A2
UniProt ID: (Human) Q9BXI2
Entrez Gene ID: (Human) 83884, (Bovine) 539296, (Mouse) 83885, (Rat) 291640
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.