Novus Biologicals
Manufacturer Code:NBP180528
Catalog # NBP180528
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC25A19. Peptide sequence VHRGSVYDAREFSVHFVCGGLAACMATLTVHPVDVLRTRFAAQGEPKVYN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNC MCPHA mitochondrial thiamine pyrophosphate carrier mitochondrial uncoupling protein 1 MUP1 solute carrier family 25 (mitochondrial deoxynucleotide carrier) member 19 solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier) member 19 TPC; Mitochondrial thiamine pyrophosphate carrier; Mitochondrial uncoupling protein 1; solute carrier family 25 (mitochondrial deoxynucleotide carrier), member 19; solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19; Solute carrier family 25 member 19
Gene Aliases: DNC; MCPHA; MUP1; SLC25A19; THMD3; THMD4; TPC
UniProt ID: (Human) Q9HC21
Entrez Gene ID: (Human) 60386
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.