Invitrogen
Manufacturer Code:PA566354
Catalog # PIPA566354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human SLC25A17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 34 kDa peroxisomal membrane protein; Peroxisomal membrane protein PMP34; PMP34 34 kDa peroxisomal membrane protein peroxisomal membrane protein (34kD) peroxisomal membrane protein PMP34 solute carrier family 25 member 17; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; Solute carrier family 25 member 17
Gene Aliases: PMP34; SLC25A17
UniProt ID: (Human) O43808
Entrez Gene ID: (Human) 10478
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.