Invitrogen
Manufacturer Code:PA526180
Catalog # PIPA526180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 17-44 amino acids from the N-terminal region of human SLC25A17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 34 kDa peroxisomal membrane protein; peroxisomal integral membrane protein 47; Peroxisomal membrane protein PMP34; peroxisomal membrane protein, 34 kDa; PMP34 34 kDa peroxisomal membrane protein peroxisomal membrane protein (34kD) peroxisomal membrane protein PMP34 solute carrier family 25 member 17; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; Solute carrier family 25 member 17; solute carrier family 25, member 17
Gene Aliases: 34kDa; 47kDa; PMP34; Pmp35; Pmp47; SLC25A17
UniProt ID: (Human) O43808, (Mouse) O70579
Entrez Gene ID: (Human) 10478, (Mouse) 20524
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.