Invitrogen
Manufacturer Code:PA5116600
Catalog # PIPA5116600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SLC25A17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 34 kDa peroxisomal membrane protein; 34 kDa peroxisomal membrane protein 34kDa 47kDa peroxisomal integral membrane protein 47 peroxisomal membrane protein PMP34 peroxisomal membrane protein 34 kDa Pmp34 Pmp35 Pmp47 SLC25A17 solute carrier family 25 (mitochondrial carrier peroxisomal membrane protein) member 17 solute carrier family 25 (mitochondrial carrier peroxisomal membrane protein 34kDa) member 17 solute carrier family 25 member 17 solute carrier family 25 member 17; peroxisomal integral membrane protein 47; Peroxisomal membrane protein PMP34; peroxisomal membrane protein, 34 kDa; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; Solute carrier family 25 member 17; solute carrier family 25, member 17
Gene Aliases: 34kDa; 47kDa; PMP34; Pmp35; Pmp47; SLC25A17
UniProt ID: (Human) O43808, (Mouse) O70579
Entrez Gene ID: (Human) 10478, (Mouse) 20524
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.