Abnova
Manufacturer Code:PAB5370
Catalog # 89-131-834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human SLC25A16. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC25A16.
For Research Use Only
Protein Aliases: D10S105E; GDA; GDC; Graves disease autoantigen; Graves disease carrier protein; HGT.1; hML7; MGC39851; Mitochondrial solute carrier protein homolog; ML7; solute carrier family 25 (mitochondrial carrier; Graves disease autoantigen), member 16; Solute carrier family 25 member 16
Gene Aliases: D10S105E; GDA; GDC; HGT.1; hML7; ML7; SLC25A16
UniProt ID: (Human) P16260
Entrez Gene ID: (Human) 8034
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.