Invitrogen
Manufacturer Code:PA542431
Catalog # PIPA542431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC25A14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BMCP-1; Brain mitochondrial carrier protein 1; Mitochondrial uncoupling protein 5; Solute carrier family 25 (mitochondrial carrier brain) member 14 UCP5 BMCP1; solute carrier family 25 (mitochondrial carrier, brain), member 14; Solute carrier family 25 member 14; UCP 5; uncoupling protein 5
Gene Aliases: BMCP-1; BMCP1; SLC25A14; UCP5; UCP5L; UCP5S; UNQ791/PRO1682; zgc:55596
UniProt ID: (Human) O95258, (Mouse) Q9Z2B2
Entrez Gene ID: (Horse) 100054503, (Human) 9016, (Dog) 612910, (Rabbit) 100348432, (Sheep) 100862656, (Bovine) 513415, (Guinea pig) 100734865, (Mouse) 20523, (Rat) 85263, (Zebrafish) 393133
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.