Abnova
Manufacturer Code:H00009016K
Catalog # 89-201-043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC25A14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLC25A14 peptide using ARM Technology.
For Research Use Only
Protein Aliases: BMCP-1; BMCP1 MGC149543 UCP5; Brain mitochondrial carrier protein 1; Mitochondrial uncoupling protein 5; solute carrier family 25 (mitochondrial carrier, brain), member 14; Solute carrier family 25 member 14; UCP 5
Gene Aliases: BMCP1; SLC25A14; UCP5; UNQ791/PRO1682
UniProt ID: (Human) O95258
Entrez Gene ID: (Human) 9016
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.