Invitrogen
Manufacturer Code:PA576838
Catalog # PIPA576838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SLC25A13. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARALAR2 Calcium binding mitochondrial carrier protein Aralar2 Calcium-binding mitochondrial carrier protein Aralar2 Citrin CMC2&lowbarHUMAN CTLN2 Ctrn Mitochondrial aspartate glutamate carrier 2 RGD1565889 Slc25a13 Solute carrier family 25 (citrin) member 13 Solute carrier family 25 member 13 (citrin) Solute carrier family 25 member 13; Calcium-binding mitochondrial carrier protein Aralar2; Citrin; Mitochondrial aspartate glutamate carrier 2; solute carrier family 25 (aspartate/glutamate carrier), member 13; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13; Solute carrier family 25 member 13; solute carrier family 25, member 13; solute carrier family 25, member 13 (citrin)
Gene Aliases: AI785475; ARALAR2; CITRIN; CTLN2; Ctrn; RGD1565889; SLC25A13
UniProt ID: (Human) O14575, (Mouse) Q9QXX4
Entrez Gene ID: (Human) 10165, (Mouse) 50799, (Rat) 362322
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.